Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPAR gamma Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120040 Supplier Invitrogen™ Supplier No. PA5120040
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: PC-3, Mouse liver, Rat fat. The target is usually found in the following locations: Cytoplasm, Nucleus. Immunogen sequence: YYSEKTQLYN KPHE.

This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR gamma
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PPARG
Gene Accession No. O88275, P37231, P37238
Gene Alias CIMT1; GLM1; HUMPPARG; NR1C3; nuclear hormone receptor; nuclear hormone receptor; PPARG; Nuclear receptor subfamily 1 group C member 3; nuclear receptor transcription factor; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferative activated receptor, gamma; peroxisome proliferative activated receptor, gamma-2; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 1; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activated receptor gamma-1; peroxisome proliferator activated receptor gamma-2; peroxisome proliferator activatived receptor gamma; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated nuclear receptor gamma variant 1; peroxisome proliferator-activated receptor; peroxisome proliferator-activated receptor gamma; peroxisome proliferator-activated receptor gamma 1; peroxisome proliferator-activated receptor gamma 1a; peroxisome proliferator-activated receptor gamma 1-a; peroxisome proliferator-activated receptor gamma 1b; peroxisome proliferator-activated receptor gamma 1-b; peroxisome proliferator-activated receptor gamma 1c; peroxisome proliferator-activated receptor gamma 1d; peroxisome proliferator-activated receptor gamma 2; peroxisome proliferator-activated receptor-gamma; peroxisome proliferator-activated receptor-gamma 2; peroxisome proliferator-cctivated receptor gamma 1; PPAR gamma; PPAR gamma 1; PPAR gamma 3; Pparg; PPARG1; PPARG2; PPARgamma; PPAR-gamma; PPARgamma2; PPAR-gamma2; PPARy; proliferator activated gamma; transcription factor
Gene Symbols PPARG
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human PPAR gamma (NP_0569532).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19016, 25664, 5468
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.