Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPAR gamma Rabbit anti-Human, Clone: 45G1, Bioss

Rabbit Monoclonal Antibody
Supplier: Bioss BSM52220R

Description
This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.Specifications
PPAR gamma | |
Monoclonal | |
1 μg/mL | |
TBS with 1% BSA, 40% glycerol and 0.05% sodium azide; pH | |
P37231 | |
PPARG | |
Recombinant human PPAR gamma protein, around 100-200aa. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
45G1 | |
Unconjugated | |
PPARG | |
CIMT1; GLM1; HUMPPARG; Nr1c3; Nuclear receptor subfamily 1 group C member 3; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated nuclear receptor gamma variant 1; Peroxisome proliferator-activated receptor gamma; PPAR gamma; PPARG; PPARG1; PPARG2; PPARgamma; PPAR-gamma; PPARgamma2; PPAR-gamma2; PPARy | |
Rabbit | |
Protein A | |
RUO | |
5468 | |
-20°C | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction