Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPAR gamma Recombinant Rabbit Monoclonal Antibody (JF101-4)
GREENER_CHOICE

Catalog No. PIMA532533
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA532533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA532533 Supplier Invitrogen™ Supplier No. MA532533
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.

This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) subfamily of nuclear receptors. PPARs form heterodimers with retinoid X receptors (RXRs) and these heterodimers regulate transcription of various genes. Three subtypes of PPARs are known: PPAR-alpha, PPAR-delta, and PPAR-gamma. The protein encoded by this gene is PPAR-gamma and is a regulator of adipocyte differentiation. Additionally, PPAR-gamma has been implicated in the pathology of numerous diseases including obesity, diabetes, atherosclerosis and cancer. Alternatively spliced transcript variants that encode different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAR gamma
Applications Western Blot
Classification Recombinant Monoclonal
Clone JF101-4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene PPARG
Gene Accession No. O88275, P37231, P37238
Gene Alias CIMT1; GLM1; HUMPPARG; NR1C3; nuclear hormone receptor; nuclear hormone receptor; PPARG; Nuclear receptor subfamily 1 group C member 3; nuclear receptor transcription factor; OTTHUMP00000185032; OTTHUMP00000185036; peroxisome proliferative activated receptor, gamma; peroxisome proliferative activated receptor, gamma-2; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor gamma; peroxisome proliferator activated receptor gamma 1; peroxisome proliferator activated receptor gamma 2; peroxisome proliferator activated receptor gamma 4; peroxisome proliferator activated receptor gamma-1; peroxisome proliferator activated receptor gamma-2; peroxisome proliferator activatived receptor gamma; peroxisome proliferator activator receptor, gamma; peroxisome proliferator-activated nuclear receptor gamma variant 1; peroxisome proliferator-activated receptor; peroxisome proliferator-activated receptor gamma; peroxisome proliferator-activated receptor gamma 1; peroxisome proliferator-activated receptor gamma 1a; peroxisome proliferator-activated receptor gamma 1-a; peroxisome proliferator-activated receptor gamma 1b; peroxisome proliferator-activated receptor gamma 1-b; peroxisome proliferator-activated receptor gamma 1c; peroxisome proliferator-activated receptor gamma 1d; peroxisome proliferator-activated receptor gamma 2; peroxisome proliferator-activated receptor-gamma; peroxisome proliferator-activated receptor-gamma 2; peroxisome proliferator-cctivated receptor gamma 1; PPAR gamma; PPAR gamma 1; PPAR gamma 3; Pparg; PPARG1; PPARG2; PPARgamma; PPAR-gamma; PPARgamma2; PPAR-gamma2; PPARy; proliferator activated gamma; transcription factor
Gene Symbols PPARG
Host Species Rabbit
Immunogen Synthetic peptide within Human PPAR gamma aa 107-150.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19016, 25664, 5468
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.