Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPARA Monoclonal Antibody (OTI2G6), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA503545S


Description
PPARA Monoclonal antibody specifically detects PPARA in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
PPARA | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q07869 | |
Mouse | |
Affinity chromatography | |
RUO | |
5465 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI2G6 | |
Unconjugated | |
PPARA | |
4933429D07Rik; AW742785; hPPAR; I79_022392; MGC2237; MGC2452; Nr1c1; nuclear hormone receptor; nuclear receptor; nuclear receptor subfamily 1 group C member 1; peroxisome proliferative activated receptor alpha; peroxisome proliferative activated receptor, alpha; peroxisome proliferative activated receptor, alpha isoform 1; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor alpha; peroxisome proliferator-activated nuclear receptor alpha variant 3; peroxisome proliferator-activated receptor alpha; PPAR; PPAR ALPHA; Ppara; PPARALPHA; PPAR-alpha; proliferator-activated receptor alpha; transcription factor belonging to the nuclear receptor superfamily | |
Human recombinant protein fragment corresponding to amino acids 145-420 of human PPARA produced in E.coli. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction