Learn More
PPARD Mouse anti-Human, Clone: OTI4D10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF811238

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the peroxisome proliferator-activated receptor (PPAR) family. PPARs are nuclear hormone receptors that bind peroxisome proliferators and control the size and number of peroxisomes produced by cells. PPARs mediate a variety of biological processes, and may be involved in the development of several chronic diseases, including diabetes, obesity, atherosclerosis, and cancer. This protein is a potent inhibitor of ligand-induced transcription activity of PPAR alpha and PPAR gamma. It may function as an integrator of transcription repression and nuclear receptor signaling. The expression of this gene is found to be elevated in colorectal cancer cells. The elevated expression can be repressed by adenomatosis polyposis coli (APC), a tumor suppressor protein related to APC/beta-catenin signaling pathway. Knockout studies in mice suggested the role of this protein in myelination of the corpus callosum, lipid metabolism, and epidermal cell proliferation.Specifications
PPARD | |
Monoclonal | |
Unconjugated | |
PPARD | |
FAAR, NR1C2, NUC1, NUCI, NUCII, PPARB | |
Mouse | |
Affinity Chromatography | |
RUO | |
5467 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4D10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q03181 | |
PPARD | |
Human recombit protein fragment corresponding to amino acids 1-244 of human PPARD produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.