Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPARd Polyclonal Antibody, Abbomax

Catalog No. 502203872
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-3872 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-3872 Supplier Abbomax Inc Supplier No. 600180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: HT-29 Cellular location: Nuclear.

PPARd Polyclonal antibody specifically detects PPARd in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen PPARd
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with proprietary stabilizer and 0.01% sodium azide
Gene Ppard
Gene Accession No. Q03181
Gene Alias FAAR; MGC3931; Nr1c2; NUC1; NUC-1; NUCI; NUCII; nuclear hormone receptor; nuclear hormone receptor 1; Nuclear receptor subfamily 1 group C member 2; OTTHUMP00000016256; OTTHUMP00000016257; peroxisome proliferative activated receptor delta; peroxisome proliferator; peroxisome proliferator activated receptor; peroxisome proliferator activated receptor beta; peroxisome proliferator activated receptor delta; peroxisome proliferator activator receptor beta; peroxisome proliferator activator receptor delta; peroxisome proliferator activator receptor, delta; peroxisome proliferator-activated nuclear receptor beta/delta variant 2; Peroxisome proliferator-activated receptor beta; Peroxisome proliferator-activated receptor delta; PPAR[b]; PPARB; Pparb/d; PPARbeta; PPAR-beta; Ppard; PPARdelta; PPAR-delta; PPARdelta/beta
Host Species Rabbit
Immunogen The full-length recombinant protein of human PPAR-delta
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5467
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.