Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPARGC1B Monoclonal Antibody (6C3F6)
GREENER_CHOICE

Catalog No. PIMA531858
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531858 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531858 Supplier Invitrogen™ Supplier No. MA531858
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-31858 has been tested in indirect ELISA.

PGC-1 beta is a tissue specific co-activator that enhances the activity of estrogen receptor alpha. The co-activation function relies on a bipartite transcriptional activation domain and two LXXLL motifs that interact with the AR2 domain of ER-alpha in an estrogen-dependent manner. Ectopic expression of this protein has been shown to lead to increased mitochondrial number and basal oxygen consumption.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPARGC1B
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6C3F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Ppargc1b
Gene Accession No. Q86YN6
Gene Alias 4631412G21Rik; ERR ligand 1; ERRL1; Perc; peroxisome proliferative activated receptor gamma coactivator 1 beta; peroxisome proliferative activated receptor, gamma, coactivator 1 beta; peroxisome proliferator-activated receptor gamma coactivator 1 beta; peroxisome proliferator-activated receptor gamma coactivator 1-beta; peroxisome proliferator-activated receptor gamma coactivator 1beta-2a; peroxisome proliferator-activated receptor gamma, coactivator 1 beta; PGC1; pgc-1 beta; PGC-1(beta); Pgc1b; PGC1beta; PGC-1beta; PGC-1-beta; PGC-1beta/ERRL1; PGC-1-related estrogen receptor alpha coactivator; PPAR gamma coactivator-1beta protein; PPARG coactivator 1 beta; PPARgamma coactivator 1 beta; PPAR-gamma coactivator 1-beta; PPARGC1; PPARGC1B; PPARGC-1-beta
Gene Symbols Ppargc1b
Host Species Mouse
Immunogen Purified recombinant fragment of human PPARGC1B (AA: 195-414) expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 133522
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.