Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPAT Mouse anti-Human, Mouse, Clone: OTI1B2, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA504559

Description
The protein encoded by this gene is a member of the purine/pyrimidine phosphoribosyltransferase family. This protein is a regulatory allosteric enzyme that catalyzes the first step of de novo purine nucleotide biosynthesis. This gene and PAICS/AIRC, a bifunctional enzyme catalyzing steps six and seven in the purine nucleotide biosynthesis pathway, are located in close proximity on chromosome 4.Specifications
PPAT | |
Monoclonal | |
0.68 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q06203 | |
Ppat | |
Human recombit protein fragment corresponding to amino acids 42-278 of human PPAT produced in E.coli. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI1B2 | |
Unconjugated | |
PPAT | |
ATASE, GPAT, PRAT | |
Mouse | |
Affinity Chromatography | |
RUO | |
231327, 5471 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction