Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550419
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550419 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550419 Supplier Invitrogen™ Supplier No. PA550419
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PPAT protein.

The protein encoded by this gene is a member of the purine/pyrimidine phosphoribosyltransferase family. This protein is a regulatory allosteric enzyme that catalyzes the first step of de novo purine nucleotide biosynthesis. This gene and PAICS/AIRC, a bifunctional enzyme catalyzing steps six and seven in the purine nucleotide biosynthesis pathway, are located in close proximity on chromosome 4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPAT
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 2.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Ppat
Gene Accession No. Q06203
Gene Alias 5730454C12Rik; AA408689; AA675351; AI507113; Amidophosphoribosyltransferase; amidophosphoribosyltransferase proprotein; amidophosphoribosyltransferase-like protein; ATASE; C79945; EC 2.4.2.14; EGK_15868; glutamine phosphoribosylpyrophosphatate amidotransferase; glutamine phosphoribosylpyrophosphate amidotransferase; glutamine PRPP amidotransferase; GPAT; phosphoribosyl pyrophosphate amidotransferase; PPAT; PRAT
Gene Symbols Ppat
Host Species Rabbit
Immunogen Fusion protein corresponding to a region derived from internal residues of human phosphoribosyl pyrophosphate amidotransferase.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 231327, 5471
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.