Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPBP/NAP2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 133131AP150UL
Description
LA-PF4 stimulates DNA synthesis. mitosis. glycolysis. intracellular cAMP accumulation. prostaglandin E2 secretion. and synthesis of hyaluronic acid and sulfated glycosaminoglycan. It also stimulates the formation and secretion of plasminogen activator bySpecifications
| PPBP/NAP2 | |
| Polyclonal | |
| Unconjugated | |
| PPBP | |
| B TG1, Beta TG, C X C motif chemokine 7, CTAP III, CTAP3, CTAPIII, CXCL7, LA PF4, LDGF, MDGF, NAP 2, NAP 2(1 63), NAP-2, PBP, Platelet basic protein, PPBP, SCYB7, Small inducible cytokine B7, TC1, TC2, TGB, TGB1, THBGB, THBGB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5473, 57349 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P02775, P09242 | |
| PPBP | |
| PPBP,NAP2 Fusion Protein Ag4147 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction