Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech PPBP Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. p-7245673
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P03BT25 25 μg
500P03BT50 50 μg
50-271-8726 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P03BT25 Supplier PeproTech Supplier No. 500P03BT25UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: AELRCMCIKT TSGIHPKNIQ SLEVIGKGTH CNQVEVIATL KDGRKICLDP DAPRIKKIVQ KKLAGDESAD. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human NAP-2 (CXCL7). Anti-Human NAP-2 (CXCL7)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human NAP-2 by sandwich ELISA (using 100 μL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human NAP-2 (CXCL7) (500-P03) as a capture antibody, allows the detection of at least 2000-4000 pg/mL of Recombinant Human NAP-2. Western Blot: To detect Human NAP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. When used in conjunction with compatible development reagents the detection limit for Recombinant Human NAP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P03BT-1MG will be provided as 2 x 500 μg

Members of the a-chemokine subfamily of inducible, secreted, pro-inflammatory cytokines contain a similar motif, in which the first two cysteine residues are separated by a single residue (Cys-X-Cys), and are also chemotactic for neutrophils. The platelet basic protein (PBP), a member of the a-chemokine family, resides in the a-granules of platelets and is released upon their activation. Proteolytic cleavage of the amino terminus of PBP leads to the generation of several peptides, which include mature PBP, connective tissue-activating peptide III (CTAP III, also designated low affinity platelet factor IV (LA-PF4)), b-thromboglobulin (b-TG), and neutrophil-activating peptide 2 (NAP-2). PBP and its N-truncated derivatives mediate inflammation and wound healing. Specifically, NAP-2 activates chemotaxis and degranulation in neutrophils during inflammation. The gene encoding human PBP maps to chromosome 4q12-q13.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPBP
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene PPBP
Gene Accession No. P02775
Gene Alias 2400003M24Rik; AI854500; Beta-TG; Beta-thromboglobulin; b-TG1; chemokine (C-X-C motif) ligand 7; connective tissue-activating peptide III; Connective tissue-activating peptide III(1-81); CTAP3; CTAPIII; CTAP-III; CTAP-III(1-81); CXC chemokine ligand 7; C-X-C motif chemokine 7; CXCL7; LA-PF4; LDGF; Leukocyte-derived growth factor; low-affinity platelet factor IV; Macrophage-derived growth factor; MDGF; NAP-2; NAP-2(1-63); NAP-2(1-66); NAP-2(73); NAP-2(74); NAP-2-L1; neutrophil activating peptide-2; neutrophil-activating peptide 2; Neutrophil-activating peptide 2(1-63); Neutrophil-activating peptide 2(1-66); Neutrophil-activating peptide 2(73); Neutrophil-activating peptide 2(74); neutrophil-activating peptide-2; PBP; Platelet basic protein; PPBP; pro-platelet basic protein; pro-platelet basic protein (chemokine (C-X-C motif) ligand 7); SCYB7; small inducible cytokine B7; small inducible cytokine subfamily B, member 7; small-inducible cytokine B7; TC1; TC-1; TC2; TC-2; TGB; TGB1; THBGB; THBGB1; thrombocidin 1; thrombocidin 2; thromboglobulin, beta-1
Gene Symbols PPBP
Host Species Rabbit
Immunogen E.coli-derived, 7.6 kDa Recombinant Human NAP-2 (CXCL7).
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5473
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.