Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPCDC Antibody (008), Alexa Fluor™ 594, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290293AF594
Description
PPCDC Monoclonal specifically detects PPCDC in Human samples. It is validated for ELISA.Specifications
PPCDC | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
8 | |
ELISA | |
coaC, EC 4.1.1.36, FLJ14585, MDS018, phosphopantothenoylcysteine decarboxylase, PPC-DC | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human PPCDC (AAH14409.1; Met1-Ser204). | |
0.1 mL | |
metabolism | |
60490 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction