Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPCDC Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300333PE
Description
PPCDC Polyclonal antibody specifically detects PPCDC in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
PPCDC | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
coaC, EC 4.1.1.36, FLJ14585, MDS018, phosphopantothenoylcysteine decarboxylase, PPC-DC | |
Produced in rabbits immunized with purified, recombinant Human PPCDC (Accession#: AAH14409.1; Met1-Ser204) | |
0.1 mL | |
metabolism | |
60490 | |
Store at 4C in the dark. | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction