Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPCDC Monoclonal Antibody (03)
GREENER_CHOICE

Catalog No. PIMA530775
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530775 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530775 Supplier Thermo Scientific Supplier No. MA530775
Add to Cart
Add to Cart

PPCDC Monoclonal Antibody (3)

PPCDC Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen PPCDC
Applications ELISA
Classification Monoclonal
Clone 03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene PPCDC
Gene Accession No. Q96CD2
Gene Alias 1810057I13Rik; 8430432M10Rik; Coac; hypothetical protein MDS018; MDS018; Phosphopantothenoylcysteine decarboxylase; Ppcdc; PPC-DC; RGD1306267; UNQ9365/PRO34154
Gene Symbols PPCDC
Host Species Mouse
Immunogen Recombinant Human PPC-DC/PPCDC Protein (Met1-Ser204).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60490
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.