Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPCDC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561065
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561065 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561065 Supplier Invitrogen™ Supplier No. PA561065
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLVAPLDANT LGKVASGICD NLLTCVMRAW DRSKPLLFCP AMNTAMWEHP ITAQQVDQLK AFGYVEIPCV AKKLVCGDEG LGAMAEVGTI V Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 88%.

Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. PPCDC, one of the last enzymes in this pathway, converts phosphopantothenoylcysteine to 4-prime-phosphopantetheine.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPCDC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPCDC
Gene Accession No. Q96CD2
Gene Alias 1810057I13Rik; 8430432M10Rik; Coac; hypothetical protein MDS018; MDS018; Phosphopantothenoylcysteine decarboxylase; Ppcdc; PPC-DC; RGD1306267; UNQ9365/PRO34154
Gene Symbols PPCDC
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPCDC. Recombinant protein control fragment (Product #RP-98087).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60490
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.