Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPCS Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531620
Description
Recommended positive controls: 293T. Predicted reactivity: Mouse (85%), Rat (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Biosynthesis of coenzyme A (CoA) from pantothenic acid (vitamin B5) is an essential universal pathway in prokaryotes and eukaryotes. PPCS (EC 6.3.2.5), one of the last enzymes in this pathway, converts phosphopantothenate to phosphopantothenoylcysteine.
Specifications
PPCS | |
Polyclonal | |
Unconjugated | |
PPCS | |
6330579B17Rik; AI413471; AI790434; Coab; Phosphopantothenate--cysteine ligase; phosphopantothenoylcysteine synthetase; PPC synthetase; Ppcs; RGD1311399 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79717 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.69 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9HAB8 | |
PPCS | |
Recombinant protein encompassing a sequence within the center region of human PPCS. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction