Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HF Scientific™ PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing

Click to view available options
For Use With (Application):
Chlorine Testing
Free Chlorine Testing
Total Chlorine Testing
For Use With (Equipment):
HF Scientific, Hach
Hach, Hanna
Non-HF, Hach or Hanna
Volume (Metric):
5 mL
10 mL
Description
Portable dispenser accurately delivers the correct volume of DPD reagent for Free or Total Chlorine—use with any DPD-based photometer or test kit.
- Supplied with enough DPD reagent for 400 tests
- To use, simply invert dispenser over sample and press button to dispense reagent; powder dissolves quickly in sample, leaving no messy residue behind
- Available for 5 and 10mL sample sizes, with formulations for a variety of photometers and test kits
Specifications
Specifications
Volume (Metric) | 10 mL |
For Use With (Application) | Total Chlorine Testing |
For Use With (Equipment) | Non-HF, Hach or Hanna |
Quantity | 400 Tests |
Type | Chlorine Reagent Powder Pop Dispenser |
Volume (English) | 0.33 oz. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction