Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPEF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556980
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556980 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556980 Supplier Invitrogen™ Supplier No. PA556980
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILVIHGGISE TTDLNLLHRV ERNKMKSVLI PPTETNRDHD TDSKHNKVGV TFNAHGRIKT NGSPTEHLTE HEWEQIIDIL WSDP Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 43%.

This gene encodes a member of the serine/threonine protein phosphatase with EF-hand motif family. The protein contains a protein phosphatase catalytic domain, and at least two EF-hand calcium-binding motifs in its C terminus. Although its substrate(s) is unknown, the encoded protein has been suggested to play a role in specific sensory neuron function and/or development. This gene shares high sequence similarity with the Drosophila retinal degeneration C (rdgC) gene. Several alternatively spliced transcript variants, each encoding a distinct isoform, have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPEF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPEF1
Gene Accession No. O14829
Gene Alias DRES10; PP7; Ppef; PPEF 1; PPEF1; PPEF-1; PPP7C; PPP7CA; protein phosphatase 7, catalytic subunit, alpha isozyme; protein phosphatase with EF calcium-binding domain; protein phosphatase with EF hand calcium-binding domain 1; protein phosphatase with EF-hand domain 1; protein phosphatase, EF-hand calcium binding domain 1; protein phosphatase, serine/threonine type, with EF-hands; RP3-499B10.1; serine/threonine protein phosphatase 7; serine/threonine-protein phosphatase 7; Serine/threonine-protein phosphatase with EF-hands 1
Gene Symbols PPEF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPEF1. Recombinant protein control fragment (Product #RP-95431).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5475
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.