Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPEF2 Polyclonal Antibody

Catalog No. PIPA535955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535955 Supplier Invitrogen™ Supplier No. PA535955
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (87%), Rat (88%), Bovine (87%).

This gene encodes a member of the serine/threonine protein phosphatase with EF-hand motif family. The protein contains a protein phosphatase catalytic domain, and at least two EF-hand calcium-binding motifs in its C terminus. Although its substrate(s) is unknown, the encoded protein, which is expressed specifically in photoreceptors and the pineal, has been suggested to play a role in the visual system. This gene shares high sequence similarity with the Drosophila retinal degeneration C gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPEF2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Ppef2
Gene Accession No. O14830, O35385
Gene Alias Ppef2; PPEF-2; PPP7CB; protein phosphatase 7, catalytic subunit, beta isozyme; protein phosphatase with EF hands 2; protein phosphatase with EF-hand domain 2; protein phosphatase, EF hand calcium-binding domain 2; protein phosphatase, EF-hand calcium binding domain 2; serine/threonine-protein phosphatase with EF-hands 2
Gene Symbols Ppef2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PPEF2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19023, 5470
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.