Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPFIA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119372
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119372 200 μL
PIPA5121672 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119372 Supplier Invitrogen™ Supplier No. PA5119372
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PPFIA1 Polyclonal Antibody for Western Blot, ICC/IF

PPFIA1 may regulate the disassembly of focal adhesions. May localize receptor-like tyrosine phosphatases type 2A at specific sites on the plasma membrane, possibly regulating their interaction with the extracellular environment and their association with substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPFIA1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Ppfia1
Gene Accession No. Q13136
Gene Alias C030014K08Rik; C87158; FLJ41337; FLJ42630; FLJ43474; LAR-interacting protein 1; LIP.1; LIP1; LIP-1; LIPRIN; Liprin-alpha1; liprin-alpha-1; MGC26800; PPFIA1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein, alpha 1; PTPRF interacting protein alpha 1; PTPRF-interacting protein alpha-1
Gene Symbols Ppfia1
Host Species Rabbit
Immunogen Recombinant protein Tyrosine Phosphatase F Interacting Protein 1. The antigen corresponds to amino acid range 1-250 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8500
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.