Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPFIA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100013
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100013 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100013 Supplier Invitrogen™ Supplier No. PA5100013
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Liprin alpha 1.

PPFIA1 may regulate the disassembly of focal adhesions. May localize receptor-like tyrosine phosphatases type 2A at specific sites on the plasma membrane, possibly regulating their interaction with the extracellular environment and their association with substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPFIA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ppfia1
Gene Accession No. Q13136
Gene Alias C030014K08Rik; C87158; FLJ41337; FLJ42630; FLJ43474; LAR-interacting protein 1; LIP.1; LIP1; LIP-1; LIPRIN; Liprin-alpha1; liprin-alpha-1; MGC26800; PPFIA1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein, alpha 1; PTPRF interacting protein alpha 1; PTPRF-interacting protein alpha-1
Gene Symbols Ppfia1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPFIA1(Accession Q13136), corresponding to amino acid residues R1150-Y1200.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 233977, 293645, 8500
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.