Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPFIA1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA522375

Catalog No. PIPA522375


Only null left
Add to Cart

Description

Description

Recommended positive controls: 293T, A431, HeLa, HepG2, JurKat, Neuro2A. Predicted reactivity: Mouse (96%), Rat (95%), Zebrafish (83%), Xenopus laevis (86%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PPFIA1 may regulate the disassembly of focal adhesions. May localize receptor-like tyrosine phosphatases type 2A at specific sites on the plasma membrane, possibly regulating their interaction with the extracellular environment and their association with substrates.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PPFIA1
Polyclonal
Unconjugated
Ppfia1
C030014K08Rik; C87158; FLJ41337; FLJ42630; FLJ43474; LAR-interacting protein 1; LIP.1; LIP1; LIP-1; LIPRIN; Liprin-alpha1; liprin-alpha-1; MGC26800; PPFIA1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 1; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein, alpha 1; PTPRF interacting protein alpha 1; PTPRF-interacting protein alpha-1
Rabbit
Antigen affinity chromatography
RUO
233977, 293645, 8500
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry
0.98 mg/mL
PBS with 20% glycerol; pH 7
Q13136
Ppfia1
Recombinant protein encompassing a sequence within the N-terminus region of human Liprin alpha 1. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.