Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPFIA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531450
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531450 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531450 Supplier Invitrogen™ Supplier No. PA531450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain. Predicted reactivity: Mouse (98%), Rat (98%), Zebrafish (85%), Rhesus Monkey (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is a member of the LAR protein-tyrosine phosphatase-interacting protein (liprin) family. Liprins interact with members of LAR family of transmembrane protein tyrosine phosphatases, which are known to be important for axon guidance and mammary gland development. It has been proposed that liprins are multivalent proteins that form complex structures and act as scaffolds for the recruitment and anchoring of LAR family of tyrosine phosphatases. This protein is most closely related to PPFIA1, a liprin family member known to interact with the protein phosphatase LAR. The expression of this gene is found to be downregulated by androgens in a prostate cancer cell line.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPFIA2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PPFIA2
Gene Accession No. O75334, Q8BSS9
Gene Alias 5330438O12; B230207K17Rik; E130120L08Rik; liprin-alpha 2; liprin-alpha-2; mKIAA4112; Ppfia2; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-2; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 2; PTPRF interacting protein alpha 2; PTPRF-interacting protein alpha-2
Gene Symbols PPFIA2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Liprin alpha 2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 327814, 362876, 8499
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.