Learn More
Invitrogen™ PPFIA2 Polyclonal Antibody

Description
PPFIA2 Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | PPFIA2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PPFIA2 |
| Gene Accession No. | O75334, Q8BSS9 |
| Gene Alias | 5330438O12; B230207K17Rik; E130120L08Rik; liprin-alpha 2; liprin-alpha-2; mKIAA4112; Ppfia2; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-2; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 2; PTPRF interacting protein alpha 2; PTPRF-interacting protein alpha-2 |
| Gene Symbols | PPFIA2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.