Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPFIA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA538012
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA538012 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA538012 Supplier Invitrogen™ Supplier No. PA538012
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A recommended positive control is 293 cell lysate.

The PPFIA3 protein is a member of the LAR protein-tyrosine phosphatase-interacting protein (liprin) family, a family of transmembrane protein tyrosine phosphatases known to be important for axon guidance and mammary gland development. Similar to the other liprin family members, PPFIA3 can interact with ERC2, a member of the ELKS-Rab6-interacting protein-CAST family of proteins at the presynaptic termini, suggesting that these proteins may play a role in the molecular organization of presynaptic zones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPFIA3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PPFIA3
Gene Accession No. O75145, P60469, Q91Z79
Gene Alias 2410127E16Rik; KIAA0654; liprin; liprin-alpha 3; Liprin-alpha3; liprin-alpha-3; liprin-alpha-3; LOW QUALITY PROTEIN: liprin-alpha-3; LPNA3; PPFIA3; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-3; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 3; protein tyrosine phosphatase, receptor type, f polypeptide, alpha 3; PTPRF interacting protein alpha 3; PTPRF-interacting protein alpha-3
Gene Symbols PPFIA3
Host Species Rabbit
Immunogen 17 amino acid peptide near the carboxy terminus of human PPFIA3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140591, 76787, 8541
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.