Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPFIA3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562173
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562173 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562173 Supplier Invitrogen™ Supplier No. PA562173
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QDELLLNKEQ LLAEMERMQM EIDQLRGRPP SSYSRSLPGS ALELRYSQAP TLPSGAHLDP YVAGSGRAGK RGRWSGVKEE PSKDWERSA Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

The PPFIA3 protein is a member of the LAR protein-tyrosine phosphatase-interacting protein (liprin) family, a family of transmembrane protein tyrosine phosphatases known to be important for axon guidance and mammary gland development. Similar to the other liprin family members, PPFIA3 can interact with ERC2, a member of the ELKS-Rab6-interacting protein-CAST family of proteins at the presynaptic termini, suggesting that these proteins may play a role in the molecular organization of presynaptic zones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPFIA3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPFIA3
Gene Accession No. O75145
Gene Alias 2410127E16Rik; KIAA0654; liprin; liprin-alpha 3; Liprin-alpha3; liprin-alpha-3; liprin-alpha-3; LOW QUALITY PROTEIN: liprin-alpha-3; LPNA3; PPFIA3; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-3; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 3; protein tyrosine phosphatase, receptor type, f polypeptide, alpha 3; PTPRF interacting protein alpha 3; PTPRF-interacting protein alpha-3
Gene Symbols PPFIA3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPFIA3. Recombinant protein control fragment (Product #RP-99163).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8541
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.