Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPFIA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562928
Description
Immunogen sequence: PFVDGVHSRS HMGSAADVRF SLGTTTHAPP GVHRRYSALR EESAKDWETS PLPGMLAPAA GPAFDSDPEI SDVDEDEPGG LVGSADVVS Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 77%.
PPFIA4, or liprin-alpha-4, belongs to the liprin-alpha gene family. See liprin-alpha-1 (LIP1, or PPFIA1; MIM 611054) for background on liprins.
Specifications
PPFIA4 | |
Polyclonal | |
Unconjugated | |
PPFIA4 | |
1110008G13Rik; AI448359; AI852265; C81506; KIAA0897; Liprin-alpha4; Liprin-alpha-4; LOW QUALITY PROTEIN: liprin-alpha-4; PPFIA4; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein alpha-4; protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 4; PTPRF interacting protein alpha 4; PTPRF-interacting protein alpha-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8497 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75335 | |
PPFIA4 | |
Recombinant protein corresponding to Human PPFIA4. Recombinant protein control fragment (Product #RP-100958). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction