Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPFIBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101985 Supplier Thermo Scientific Supplier No. PA5101985
Add to Cart
Add to Cart

PPFIBP1 Polyclonal Antibody

PPFIBP1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen PPFIBP1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PPFIBP1
Gene Accession No. Q86W92, Q8C8U0
Gene Alias 4632409B19Rik; AW214454; AW261454; hSGT2; hSgt2p; KIAA1230; L2; liprin beta 1; liprin related protein; liprin-beta 1; liprin-beta-1; PPFIA binding protein 1; Ppfibp1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein-binding protein 1; protein-tyrosine phosphatase receptor-type f polypeptide-interacting protein-binding protein 1; PTPRF interacting protein, binding protein 1 (liprin beta 1); PTPRF-interacting protein-binding protein 1; SGT2
Gene Symbols PPFIBP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PPFIBP1(Accession Q86W92), corresponding to amino acid residues V299-K349.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67533, 8496
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.