Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPFIBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551663
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551663 Supplier Invitrogen™ Supplier No. PA551663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DAQGFSDLEK SPSPTPVMGS PSCDPFNTSV PEEFHTTILQ VSIPSLLPAT VSMETSEKSK LTPKPETSFE ENDGNIILGA TVDTQLCDKL LTSSLQKSSS LGNLKKETSD GEKETIQKTS EDRA Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 67%.

The protein encoded by this gene is a member of the LAR protein-tyrosine phosphatase-interacting protein (liprin) family. Liprins interact with members of LAR family of transmembrane protein tyrosine phosphatases, which are known to be important for axon guidance and mammary gland development. It has been proposed that liprins are multivalent proteins that form complex structures and act as scaffolds for the recruitment and anchoring of LAR family of tyrosine phosphatases. This protein was found to interact with S100A4, a calcium-binding protein related to tumor invasiveness and metastasis. In vitro experiment demonstrated that the interaction inhibited the phosphorylation of this protein by protein kinase C and protein kinase CK2. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPFIBP1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPFIBP1
Gene Accession No. Q86W92
Gene Alias 4632409B19Rik; AW214454; AW261454; hSGT2; hSgt2p; KIAA1230; L2; liprin beta 1; liprin related protein; liprin-beta 1; liprin-beta-1; PPFIA binding protein 1; Ppfibp1; Protein tyrosine phosphatase receptor type f polypeptide-interacting protein-binding protein 1; protein-tyrosine phosphatase receptor-type f polypeptide-interacting protein-binding protein 1; PTPRF interacting protein, binding protein 1 (liprin beta 1); PTPRF-interacting protein-binding protein 1; SGT2
Gene Symbols PPFIBP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPFIBP1. Recombinant protein control fragment (Product #RP-102023).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8496
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.