Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPHLN1 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298653UV
Description
PPHLN1 Polyclonal specifically detects PPHLN1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
PPHLN1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
51535 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
DyLight 350 | |
Gastric cancer antigen Ga50, HSPC232, MGC48786, periphilin 1, periphilin-1 | |
Produced in rabbits immunized with E. coli-derived Human PPHLN1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction