Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPIAL4A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572156
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572156 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572156 Supplier Invitrogen™ Supplier No. PA572156
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PPIAL4A Polyclonal Antibody for Western Blot

PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPIAL4A
Applications Western Blot
Classification Polyclonal
Concentration 0.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene PPIAL4A
Gene Accession No. Q9Y536
Gene Alias chromosome 1-amplified sequence 2; chromosome one-amplified sequence 2; COAS2; COAS-2; Cyclophilin homolog overexpressed in liver cancer; cyclophilin-LC (COAS2); peptidyl-prolyl cis-trans isomerase A-like 4A; peptidylprolyl cis-trans isomerase A-like 4A; peptidyl-prolyl cis-trans isomerase A-like 4A/B/C; peptidylprolyl cis-trans isomerase A-like 4B; peptidylprolyl isomerase A (cyclophilin A)-like 4; peptidylprolyl isomerase A (cyclophilin A)-like 4A; peptidylprolyl isomerase A (cyclophilin A)-like 4B; peptidylprolyl isomerase A like 4A; PPIAL4; PPIAL4A; PPIAL4B; PPIAL4G; PPIase A-like 4A; PPIase A-like 4B; PPIase A-like 4G
Gene Symbols PPIAL4A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PPIAL4A.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653505
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.