Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPIAL4G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571238
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571238 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571238 Supplier Thermo Scientific Supplier No. PA571238
Add to Cart
Add to Cart

PPIAL4G Polyclonal Antibody

PPIAL4G Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PPIAL4G
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PPIAL4G
Gene Accession No. P0DN37
Gene Alias chromosome 1-amplified sequence 2; COAS-2; Peptidylprolyl cis-trans isomerase A-like 4; peptidylprolyl cis-trans isomerase A-like 4A; peptidyl-prolyl cis-trans isomerase A-like 4G; peptidylprolyl cis-trans isomerase A-like 4G; peptidylprolyl isomerase A (cyclophilin A)-like 4G; peptidylprolyl isomerase A like 4G; PPIAL4G; PPIase A-like 4A; PPIase A-like 4G; RGD1559682; similar to peptidylprolyl isomerase A (cyclophilin A))
Gene Symbols PPIAL4G
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human PPIAL4G.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644591
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.