Learn More
PPID Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305334A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein has been shown to possess PPIase activity and, similar to other family members, can bind to the immunosuppressant cyclosporin A.Specifications
PPID | |
Polyclonal | |
Unconjugated | |
PPID | |
Cyp D, Peptidyl-prolyl cis-trans isomerase D, EC 5.2.1.8, PPIase, Rotamase, S-cyclophilin, SCYLP, CYP-S1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5481, 67738 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q08752, Q9CR16 | |
Ppid | |
Between 175 and 225 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.