Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPIG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517016 Supplier Invitrogen™ Supplier No. PA517016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

Structural Maintenance of Chromosomes (SMC) family proteins play critical roles in various nuclear events that require structural changes of chromosomes, including mitotic chromosome organization, DNA recombination and repair and global transcriptional repression. The chromo proteins are conserved in eukaryotes and lead to mitotic chromosome segregation defects. This suggests a critical function of SMC family proteins in mitotic chromosome dynamics. SMC1 and SMC3 form a heterodimeric complex required for metaphase progression in mitotic cells. Specifically this SMC1/SMC3 complex is responsible for sister chromatid cohesion during metaphase. Mutations in the gene can lead to Cornelia de Lange syndrome 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPIG
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene PPIG
Gene Accession No. A2AR02, O55035, Q13427
Gene Alias AU019516; AU022200; B230312B02Rik; CARS-cyclophilin; CARS-Cyp; CASP10; Clk-associating RS-cyclophilin; Cyclophilin G; CYP; Matrin cyclophilin; matrin cyclophilin (matrin-cyp); matrin-cyp; MGC133241; OTTHUMP00000204761; OTTHUMP00000204762; Peptidyl-prolyl cis-trans isomerase G; peptidylprolyl isomerase G; peptidyl-prolyl isomerase G; peptidylprolyl isomerase G (cyclophilin G); peptidyl-prolyl isomerase G (cyclophilin G); PPIase G; PPIG; Rotamase G; SCAF10; SR-cyclophilin; SRCyp; SR-cyp; SR-related CTD-associated factor 10
Gene Symbols PPIG
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Glu391 of human PPIG.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228005, 83624, 9360
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.