Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPIH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588460 Supplier Invitrogen™ Supplier No. PA588460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAVANSSPVN PVVFFDVSIG GQEVGRMKIE LFADVVPKTA ENFRQFCTGE FRKDGVPIGY KGSTFHRVIK DFMIQGGDFV NGDGTGVASI YRGPFADENF KLRHSAPGLL SMANSGPSTN GCQFFITCSK CDWLDGKHVV FGKIIDGLLV MRKIENVPTG PNNKPKLPVV ISQCGEM; Positive Samples: HepG2, Rat testis, Rat kidney; Cellular Location: Cytoplasm, Nucleus speckle.

The protein encoded by this gene is a member of the peptidyl-prolyl cis-trans isomerase (PPIase) family. PPIases catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and accelerate the folding of proteins. This protein is a specific component of the complex that includes pre-mRNA processing factors PRPF3, PRPF4, and PRPF18, as well as U4/U5/U6 tri-snRNP. This protein has been shown to possess PPIase activity and may act as a protein chaperone that mediates the interactions between different proteins inside the spliceosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPIH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PPIH
Gene Accession No. O43447
Gene Alias 1100001J08Rik; 2010111B15Rik; 4833408F11Rik; AI464484; cyclophilin H; CYP20; CYP-20; CYPH; D4Wsu43e; peptidyl prolyl isomerase H; peptidyl-prolyl cis-trans isomerase H; peptidylprolyl isomerase H; peptidylprolyl isomerase H (cyclophilin H); PPIase h; Ppih; RGD1564921; Rotamase H; small nuclear ribonucleoprotein particle-specific cyclophilin H; SnuCyp-20; USA-CYP; USA-CyP SnuCyp-20; U-snRNP-associated cyclophilin SnuCyp-20; U-snRNP-associated cyclophilin SunCyp-20
Gene Symbols PPIH
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-177 of human PPIH (NP_0063381).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10465, 366461
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.