Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPIL1 Antibody (08), FITC , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306214F
Description
PPIL1 Monoclonal antibody specifically detects PPIL1 in Human samples. It is validated for Flow Cytometry, ELISASpecifications
PPIL1 | |
Monoclonal | |
FITC | |
CYPL1cyclophilin-related gene 1, EC 5.2.1.8, hCyPX, MGC678, peptidyl-prolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 1, peptidylprolyl isomerase (cyclophilin)-like 1, PPIase, Rotamase PPIL1 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PPIL1 (Q9Y3C6; Met1-Gly166). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA | |
08 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
51645 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction