Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPIL1 Antibody (OTI2D1), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273573FR
Description
PPIL1 Monoclonal specifically detects PPIL1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PPIL1 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
PPIL1 | |
Full length recombinant protein of human PPIL1 (NP_057143) produced in E.coli. | |
0.1 mL | |
51645.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2D1 | |
Western Blot, Immunohistochemistry | |
CYPL1cyclophilin-related gene 1, EC 5.2.1.8, hCyPX, MGC678, peptidyl-prolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 1, peptidylprolyl isomerase (cyclophilin)-like 1, PPIase, Rotamase PPIL1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction