Learn More
Invitrogen™ PPIL1 Monoclonal Antibody (OTI2D1)

Description
PPIL1 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PPIL1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI2D1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | Ppil1 |
| Gene Accession No. | Q9Y3C6 |
| Gene Alias | 1110060O10Rik; AI327391; CGI-124; cyclophilin like 1; cyclophilin-related gene 1; CYPL1; hCyPX; peptidyl-prolyl cis-trans isomerase; peptidyl-prolyl cis-trans isomerase-like 1; peptidylprolyl isomerase (cyclophilin)-like 1; peptidylprolyl isomerase like 1; peptidylprolyl isomerase-like 1; PPIase; PPIL1; Rotamase PPIL1; UNQ2425/PRO4984 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.