Learn More
PPIL1 Mouse anti-Human, Clone: OTI8F8, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803795

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.Specifications
PPIL1 | |
Monoclonal | |
Unconjugated | |
PPIL1 | |
CGI-124, CYPL1, PPIase, hCyPX | |
Mouse | |
Affinity Chromatography | |
RUO | |
51645 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI8F8 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y3C6 | |
PPIL1 | |
Full length human recombit protein of human PPIL1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.