Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPIL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551300
Description
The antibody detects endogenous levels of total PPIL1 protein.
This gene is a member of the cyclophilin family of peptidylprolyl isomerases (PPIases). The cyclophilins are a highly conserved, ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. Based on similarity to other PPIases, this protein could accelerate the folding of proteins and might catalyze the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
Specifications
PPIL1 | |
Polyclonal | |
Unconjugated | |
Ppil1 | |
1110060O10Rik; AI327391; CGI-124; cyclophilin like 1; cyclophilin-related gene 1; CYPL1; hCyPX; peptidyl-prolyl cis-trans isomerase; peptidyl-prolyl cis-trans isomerase-like 1; peptidylprolyl isomerase (cyclophilin)-like 1; peptidylprolyl isomerase like 1; peptidylprolyl isomerase-like 1; PPIase; PPIL1; Rotamase PPIL1; UNQ2425/PRO4984 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51645, 68816 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.7 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q9D0W5, Q9Y3C6 | |
Ppil1 | |
Full length fusion protein of human PPIL1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction