Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PPIL2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | PPIL2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PPIL2 Polyclonal specifically detects PPIL2 in Human samples. It is validated for Western Blot.Specifications
| PPIL2 | |
| Polyclonal | |
| Rabbit | |
| Q13356 | |
| 23759 | |
| Synthetic peptides corresponding to PPIL2(peptidylprolyl isomerase (cyclophilin)-like 2) The peptide sequence was selected from the middle region of PPIL2. Peptide sequence MKIIDPDEEKAKQDPSYYLKNTNAETRETLQELYKEFKGDEILAATMKAP. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| CYC4, cyclophilin, 60kDa, cyclophilin-60, Cyclophilin-like protein Cyp-60, Cyp-60, CYP60, EC 5.2.1.8, FLJ39930, hCyP-60, MGC33174, MGC787, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 2, peptidylprolyl isomerase (cyclophilin)-like 2, PPIase, Rotamase PPIL2 | |
| PPIL2 | |
| IgG | |
| 59 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title