Learn More
Invitrogen™ PPIL2 Isoform B Recombinant Rabbit Monoclonal Antibody (015)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA530779
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human PPIL2.
This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. This protein interacts with the proteinase inhibitor eglin c and is localized in the nucleus. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
PPIL2 Isoform B | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q13356 | |
PPIL2 | |
Recombinant Human PPIL2 Protein (Gly280-Pro457). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunocytochemistry | |
015 | |
Unconjugated | |
PPIL2 | |
CYC4; cyclophilin, 60kDa; cyclophilin-60; Cyclophilin-like protein Cyp-60; CYP60; Cyp-60; hCyP-60; peptidylprolyl cis-trans isomerase; peptidyl-prolyl cis-trans isomerase-like 2; peptidylprolyl isomerase (cyclophilin)-like 2; peptidylprolyl isomerase like 2; PPIase; PPIL2; Probable inactive peptidyl-prolyl cis-trans isomerase-like 2; RING-type E3 ubiquitin-protein ligase PPIL2; Rotamase PPIL2; U-box domain containing 7; UBOX7 | |
Rabbit | |
Protein A | |
RUO | |
23759 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.