Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPIL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557237
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557237 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557237 Supplier Invitrogen™ Supplier No. PA557237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MENVESDPKT DRPKEEIRID ATTVFVDPYE EADAQIAQER KTQLKVAPET KVKSSQPQAG SQGPQTFRQG VGKYINPAAT KRAAEEEPST SATVPMSKKK PSRGFGDFSS Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 82%.

This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved ubiquitous family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions. This protein interacts with the proteinase inhibitor eglin c and is localized in the nucleus. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPIL2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPIL2
Gene Accession No. Q13356
Gene Alias 0610009L05Rik; 1700016N17Rik; 4921520K19Rik; 4930511F14Rik; AA589416; C130078A06Rik; CYC4; cyclophilin, 60kDa; cyclophilin-60; Cyclophilin-like protein Cyp-60; CYP60; Cyp-60; hCyP-60; peptidylprolyl cis-trans isomerase; peptidyl-prolyl cis-trans isomerase-like 2; peptidylprolyl isomerase (cyclophilin)-like 2; peptidylprolyl isomerase like 2; peptidylprolyl isomerase-like 2; PPIase; PPIL2; Probable inactive peptidyl-prolyl cis-trans isomerase-like 2; RING-type E3 ubiquitin-protein ligase PPIL2; Rotamase PPIL2; U-box domain containing 7; UBOX7
Gene Symbols PPIL2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PPIL2. Recombinant protein control fragment (Product #RP-95345).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23759
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.