Learn More
Invitrogen™ PPIL3 Monoclonal Antibody (OTI2E7)

Description
PPIL3 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | PPIL3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI2E7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PPIL3 |
| Gene Accession No. | Q9H2H8 |
| Gene Alias | 2310076N22Rik; 2510026K04Rik; cyclophilin J; cyclophilin-like 3; cyclophilin-like protein 3; cyclophilin-like protein PPIL3; Cyp10l; CyPJ; Peptidyl-prolyl cis-trans isomerase-like 3; peptidylprolyl cis-trans isomerase-like protein 3; peptidylprolyl isomerase (cyclophilin)-like 3; peptidylprolyl isomerase like 3; peptidylprolyl isomerase -like 3; PPIase; PPIase-like protein 3; Ppil3; Rotamase PPIL3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.