Learn More
PPIL3 Mouse anti-Human, Clone: OTI1G3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505245

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the cyclophilin family. Cyclophilins catalyze the cis-trans isomerization of peptidylprolyl imide bonds in oligopeptides. They have been proposed to act either as catalysts or as molecular chaperones in protein-folding events. Alternative splicing results in multiple transcript variants.Specifications
PPIL3 | |
Monoclonal | |
Unconjugated | |
PPIL3 | |
CYPJ | |
Mouse | |
Affinity Chromatography | |
RUO | |
53938 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Western Blot | |
OTI1G3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9H2H8 | |
PPIL3 | |
Full length human recombit protein of human PPIL3 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.