Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPIL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566465
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566465 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566465 Supplier Invitrogen™ Supplier No. PA566465
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSVTLHTDVGD IKIEVFCERT PKTCENFLAL CASNYYNGCI FHRNIKGFMV QT Highest antigen sequence identity to the following orthologs - mouse 98%, rat 98%.

This gene encodes a member of the cyclophilin family. Cyclophilins catalyze the cis-trans isomerization of peptidylprolyl imide bonds in oligopeptides. They have been proposed to act either as catalysts or as molecular chaperones in protein-folding events. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPIL3
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPIL3
Gene Accession No. Q9H2H8
Gene Alias 2310076N22Rik; 2510026K04Rik; cyclophilin J; cyclophilin-like 3; cyclophilin-like protein 3; cyclophilin-like protein PPIL3; Cyp10l; CyPJ; Peptidyl-prolyl cis-trans isomerase-like 3; peptidylprolyl cis-trans isomerase-like protein 3; peptidylprolyl isomerase (cyclophilin)-like 3; peptidylprolyl isomerase like 3; peptidylprolyl isomerase -like 3; PPIase; PPIase-like protein 3; Ppil3; Rotamase PPIL3
Gene Symbols PPIL3
Host Species Rabbit
Immunogen Recombinant Human PPIL3. Recombinant protein control fragment (Product #RP-107140).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53938
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.