Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PPIL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113337
Description
PPIL3 Polyclonal Antibody for Western Blot, IHC (P), ELISA
This gene encodes a member of the cyclophilin family. Cyclophilins catalyze the cis-trans isomerization of peptidylprolyl imide bonds in oligopeptides. They have been proposed to act either as catalysts or as molecular chaperones in protein-folding events. Alternative splicing results in multiple transcript variants.
Specifications
PPIL3 | |
Polyclonal | |
Unconjugated | |
PPIL3 | |
2310076N22Rik; 2510026K04Rik; cyclophilin J; cyclophilin-like 3; cyclophilin-like protein 3; cyclophilin-like protein PPIL3; Cyp10l; CyPJ; Peptidyl-prolyl cis-trans isomerase-like 3; peptidylprolyl cis-trans isomerase-like protein 3; peptidylprolyl isomerase (cyclophilin)-like 3; peptidylprolyl isomerase like 3; peptidylprolyl isomerase -like 3; PPIase; PPIase-like protein 3; Ppil3; Rotamase PPIL3 | |
Rabbit | |
Protein G | |
RUO | |
53938 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9H2H8 | |
PPIL3 | |
Recombinant Human Peptidyl-prolyl cis-trans isomerase-like 3 protein (2-161AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction