Learn More
Invitrogen™ PPIL4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590821
Description
Immunogen sequence: MDNVLIDDRR IHVDFSQSVA KVKWKGKGGK YTKSDFKEYE KEQDKPPNLV LKDKVKPKQD TKYDLILDEQ AEDSKSSHSH TSKKHKKKTH HCSEEKEDED YMPIKNTNQD IYREMGFGHY EEEESCWEKQ KSEKRDRTQN RSRSRSRERD GHYSNSHKSK YQTDLYERER SKKRDRSRSP KKSKDKEKSK YR; Positive Samples: 293T, U-87MG, LO2; Cellular Location: Nucleus.
This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions.
Specifications
PPIL4 | |
Polyclonal | |
Unconjugated | |
PPIL4 | |
3732410E19Rik; 3830425H19Rik; AI788954; AW146233; Cyclophilin-like protein PPIL4; cyclophilin-type peptidyl-prolyl cis-trans isomerase; HDCME13P; peptidyl-prolyl cis-trans isomerase-like 4; peptidylprolyl isomerase (cyclophilin)-like 4; peptidylprolyl isomerase like 4; peptidylprolyl isomerase-like 4; PPIase; PPIL4; Rotamase PPIL4; serologically defined breast cancer antigen NY-BR-18 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
85313 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.86 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8WUA2 | |
PPIL4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 301-492 of human PPIL4 (NP_6243111). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.