Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PPIL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111026
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111026 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111026 Supplier Invitrogen™ Supplier No. PA5111026
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSHSHTSKKH KKKTHHCSEE KEDEDYMPIK NTNQDIYREM GFGHYEEEES CWEKQKSEKR DRTQNRSRS Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 94%.

This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PPIL4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PPIL4
Gene Accession No. Q8WUA2
Gene Alias 3732410E19Rik; 3830425H19Rik; AI788954; AW146233; Cyclophilin-like protein PPIL4; cyclophilin-type peptidyl-prolyl cis-trans isomerase; HDCME13P; peptidyl-prolyl cis-trans isomerase-like 4; peptidylprolyl isomerase (cyclophilin)-like 4; peptidylprolyl isomerase like 4; peptidylprolyl isomerase-like 4; PPIase; PPIL4; Rotamase PPIL4; serologically defined breast cancer antigen NY-BR-18
Gene Symbols PPIL4
Host Species Rabbit
Immunogen Recombinant Protein corresonding to Human PPIL4. Recombinant protein control fragment (Product #RP-104638).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 85313
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.