Learn More
PPIL4 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304964AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene is a member of the cyclophilin family of peptidylprolyl isomerases. The cyclophilins are a highly conserved family, members of which play an important role in protein folding, immunosuppression by cyclosporin A, and infection of HIV-1 virions.Specifications
PPIL4 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q8WUA2, Q9CXG3 | |
Ppil4 | |
Between 325 and 375 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PPIL4 | |
HDCME13P, PPIase, cyclophilin-like protein PPIL4, cyclophilin-type peptidyl-prolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase-like 4, rotamase PPIL4, serologically defined breast cancer antigen NY-BR-18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
67418, 85313 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.